Learn More
Medchemexpress LLC Isonicotinic acid hydrazide-d4 | 774596-24-6 | 99.9% | 1 MG
SDP
Catalog No. 5000406809
Encompass_Preferred
Isoniazid-d4 is a deuterium-labeled form of Isoniazid, a proagent activated by the bacterial catalase-peroxidase enzyme KatG. It exhibits bactericidal activity against rapidly dividing mycobacteria and possesses anti-tuberculostatic properties.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06809 Supplier Medchemexpress LLC Supplier No. HYB0329S1MG
Isoniazid-d4 is a deuterium-labeled form of Isoniazid, a proagent activated by the bacterial catalase-peroxidase enzyme KatG. It exhibits bactericidal activity against rapidly dividing mycobacteria and possesses anti-tuberculostatic properties.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs