Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isookanin | 1036-49-3 | 5 MG
SDP

Catalog No. 5000417354
Encompass_Preferred

Isookanin is a compound studied for its potential in researching various illnesses including cancers, skin rashes, snake and insect bites, diabetes mellitus, and diarrhea. It functions as an anti-viral agent against HSV and varicella-zoster virus (VZV), and demonstrates antioxidant activity.

  • Purity: 99.76%
  • Molecular weight: 288.25
  • Formula: C15H12O6
  • Appearance: Solid, off-white to yellow
  • Neuroprotective activity (HT-22 EC50: 6.2 μM)
  • DPPH scavenging activity (EC50: 10.6 μM)
  • Cellular antioxidant activity (IC50: 14.4 μM)
  • Derived from plants
  • Store at 4°C (protected from light); in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Solubility in DMSO: ≥ 100 mg/mL

Catalog No. 50-004-17354 Supplier Medchemexpress LLC Supplier No. HYN76775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isookanin is a compound studied for its potential in researching various illnesses including cancers, skin rashes, snake and insect bites, diabetes mellitus, and diarrhea. It functions as an anti-viral agent against HSV and varicella-zoster virus (VZV), and demonstrates antioxidant activity.

  • Purity: 99.76%
  • Molecular weight: 288.25
  • Formula: C15H12O6
  • Appearance: Solid, off-white to yellow
  • Neuroprotective activity (HT-22 EC50: 6.2 μM)
  • DPPH scavenging activity (EC50: 10.6 μM)
  • Cellular antioxidant activity (IC50: 14.4 μM)
  • Derived from plants
  • Store at 4°C (protected from light); in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Solubility in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.