Learn More
Medchemexpress LLC HY-N0767 1mg Medchemexpress, Isoorientin CAS:4261-42-1 Purity:>98%
SDP
Catalog No. 501933706
Encompass_Preferred
Medchemexpress, HY-N0767 1mg Isoorientin CAS:4261-42-1 Isoorientin is a potent inhibitor of COX-2 with an IC 50 value of 39 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3706 Supplier Medchemexpress LLC Supplier No. HYN07671MG
Medchemexpress, HY-N0767 1mg Isoorientin CAS:4261-42-1 Isoorientin is a potent inhibitor of COX-2 with an IC 50 value of 39 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.