Learn More
Echelon Biosciences Research Labs ISOPENTENYL DIPHOSPHATE 1 MG
SDP
Supplier:  Echelon Biosciences Research Labs I00501MG
Encompass
Isopentenyl Diphosphate (tris-ammonium salt) IPP, Molecular Formula:- C5H21N3O7P2 Molecular Weight (g/mol): -297.18 Storage:-0 deg.C, or flash-frozen with liquid N2 and kept at -70 deg.C. This item is non returnable
Catalog No. 50-720-379