Learn More
Echelon Biosciences Research Labs ISOPENTENYL DIPHOSPHATE 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs I005010MG
Encompass
Isoprenoid compounds are a diverse group of natural products which are essential components in all cells. Isoprenoids are biosynthesized from the simple precursors isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). Eukaryotes, fungi, and some gram-positive bacteria produce IPP through the mevalonate (MVA) pathway whereas gram-negative and some gram-positive bacteria utilize the non-mevalonate or 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. This item is non returnable
Catalog No. 50-720-380