Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopentenyl pyrophosphate trilithium salt, ≥95.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111013397 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3397 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3397 Supplier MilliporeSigma Supelco Supplier No. 397845MG
Add to Cart
Add to Cart

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 18687-43-9
Physical Form Neat
Linear Formula C5H9 Li3O7P2
MDL Number MFCD17215950
Synonym 3-Methyl-3-butenyl pyrophosphate trilithium salt; IPP
Type Analytical Standard
Quantity 5 mg
Formula Weight 263.89
Percent Purity ≥95.0% (TLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.