Learn More
Medchemexpress LLC Isopentyl octanoate | 2035-99-6 | MFCD00048917 | 97.0% | 214.35 g/mol | C13H26O2 | 100 G
SDP
Catalog No. 5000302773
Encompass_Preferred
Isopentyl octanoate (isoamyl octanoate) is a colorless to light yellow liquid ester (C13H26O2, MW 214.35 g/mol, CAS 2035-99-6) used as a biochemical reagent and a fragrance/flavor intermediate. It is supplied in laboratory pack sizes and is characterized by low density and defined purity for analytical and formulation applications.
  • Colorless to light yellow liquid with a density of 0.861 g/cm3.
  • Suitable for use as a fragrance and flavor intermediate.
  • High purity appropriate for analytical and formulation work.
  • Compatible with common organic solvents used in lab workflows.
  • Supplied in controlled laboratory pack sizes for research use.
Catalog No. 50-003-02773 Supplier Medchemexpress LLC Supplier No. HYW014206100G
Add to Cart
Add to Cart
Isopentyl octanoate (isoamyl octanoate) is a colorless to light yellow liquid ester (C13H26O2, MW 214.35 g/mol, CAS 2035-99-6) used as a biochemical reagent and a fragrance/flavor intermediate. It is supplied in laboratory pack sizes and is characterized by low density and defined purity for analytical and formulation applications.
  • Colorless to light yellow liquid with a density of 0.861 g/cm3.
  • Suitable for use as a fragrance and flavor intermediate.
  • High purity appropriate for analytical and formulation work.
  • Compatible with common organic solvents used in lab workflows.
  • Supplied in controlled laboratory pack sizes for research use.