Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopentyl octanoate | 2035-99-6 | MFCD00048917 | 99.4% | 214.35 g/mol | C13H26O2 | 250 G
SDP

Catalog No. 5000302345
Encompass_Preferred

Isopentyl octanoate is an alkyl ester (C13H26O2) commonly used as a reagent and analytical standard in research laboratories. It is supplied as a clear liquid with measured density and high purity, making it suitable for analytical assays, chemical synthesis, and formulation studies.

  • High purity: 99.39%.
  • Clear liquid physical state.
  • Molecular weight 214.35 g/mol.
  • Density 0.861 g/cm³.
  • Suitable for analytical standards and reagent use.

Catalog No. 50-003-02345 Supplier Medchemexpress LLC Supplier No. 250GHYW014206
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopentyl octanoate is an alkyl ester (C13H26O2) commonly used as a reagent and analytical standard in research laboratories. It is supplied as a clear liquid with measured density and high purity, making it suitable for analytical assays, chemical synthesis, and formulation studies.

  • High purity: 99.39%.
  • Clear liquid physical state.
  • Molecular weight 214.35 g/mol.
  • Density 0.861 g/cm³.
  • Suitable for analytical standards and reagent use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.