Learn More
Echelon Biosciences Research Labs ISOPENTYLTHIOLO DIPHOSPHATE 10mg
SDP
Supplier:  Echelon Biosciences Research Labs IS05010MG
Encompass
Isopentenyl thiolodiphosphate (ISPP) is an analog of IPP in which the non-bridging oxygen has been replaced by sulfur resulting in a less-reactive compound. This item is non returnable
Catalog No. 50-720-408