Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isophorone diisocyanate

Catalog No. 3176245ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-762-45ML 5 mL
31-762-41L 1 L
31-762-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-762-45ML Supplier Sigma Aldrich Supplier No. 3176245ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isophorone diisocyanate (IPDI) is an aliphatic diisocyanate that is majorly used as a curing agent by forming NCO linkages. It is mainly utilized in the preparation of polyurethane based films and foams for a variety of high performance coating applications. It provides UV resistant films due to the presence of the aliphatic ring.

Specifications

Density 1.049 g/mL (at 25°C)
Boiling Point 158°C to 159°C (15 mmHg)
Molecular Formula C12H18N2O2
Refractive Index n20/D 1.484 (literature)
Linear Formula OCNC6H7(CH3)3 CH2NCO
MDL Number MFCD00064956
Quantity 5 mL
Synonym 5-Isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular Weight (g/mol) 222.28
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.