Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophorone diisocyanate, 98+%, mixture of isomers

Catalog No. AC427600010
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427600010 1 L Glass bottle
AC427600500 50 mL Glass bottle
AC427602500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC427600010 Supplier Thermo Scientific Chemicals Supplier No. 427600010
  • $318.00
    $252.65 / Each of 1

    Save $65.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-71-9
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00064956
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
PubChem CID 169132
ChEBI CHEBI:53214
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O

Specifications

Color Colorless to Yellow
Density 1.0610g/mL
Boiling Point 153.0°C (10.0 mmHg)
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (sum of mixed isomers) (GC)
Packaging Glass bottle
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Quantity 1 L
Specific Gravity 1.061
Viscosity 13-15 mPa.s (23°C)
Formula Weight 222.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isophorone diisocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic to aquatic life with long lasti
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If skin irritation or
EINECSNumber 223-861-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.