Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophorone diisocyanate, 99.3%, For GC analysis, MP Biomedicals™

Catalog No. ICN15510680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15510680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15510680 Supplier MP Biomedicals, Inc Supplier No. 0215510680
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4098-71-9
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00064956
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
PubChem CID 169132
ChEBI CHEBI:53214
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O

Specifications

Color Colorless
Density 1.058g/cm3 at 20°C
Quantity 100 g
Formula Weight 222.3
Physical Form Liquid
Chemical Name or Material Isophorone diisocyanate, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.