Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Isophthalaldehyde, 626-19-7, MFCD00003372, 25g

Catalog No. 50681168
Encompass_Preferred

Molecular Formula C8H6O2, Purity 95+%, Molecular Weight 134.14, Melting Point 89°, Boiling Point 246

Catalog No. 50-681-168 Supplier Matrix Scientific Supplier No. 07658925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Molecular Formula C8H6O2, Purity 95+%, Molecular Weight 134.14, Melting Point 89°, Boiling Point 246

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.