Learn More
Medchemexpress LLC Isophthalic-2,4,5,6 Acid-d4 | 148472-48-4 | 99.7% | 170.16 | 5 MG
SDP
Catalog No. 5000401531
Encompass_Preferred
Isophthalic-2,4,5,6 Acid-d4 is the deuterium labeled Isophthalic acid. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-01531 Supplier Medchemexpress LLC Supplier No. HY21323S5MG
Isophthalic-2,4,5,6 Acid-d4 is the deuterium labeled Isophthalic acid. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs