Learn More
Medchemexpress LLC Isophthalic-2,4,5,6 acid-d4 | 148472-48-4 | MFCD00209746 | 99.7% | 170.16 g/mol | C8H2D4O4 | 10 MG
SDP
Catalog No. 5000297763
Encompass_Preferred
Isophthalic-2,4,5,6 Acid-d4 is the deuterium-labeled analogue of isophthalic acid supplied as a solid stable-isotope standard for analytical and tracer applications. It is intended for use as an internal standard in quantitative NMR, GC-MS, and LC-MS assays and for tracer studies requiring deuterium labeling.
  • Deuterium-labeled for tracer and internal standard applications.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied as a solid powder for convenient handling and storage.
  • High chemical purity of 99.7% for reliable analytical performance.
  • Stable when stored under recommended conditions to maintain integrity.
Catalog No. 50-002-97763 Supplier Medchemexpress LLC Supplier No. HY21323S10MG
Isophthalic-2,4,5,6 Acid-d4 is the deuterium-labeled analogue of isophthalic acid supplied as a solid stable-isotope standard for analytical and tracer applications. It is intended for use as an internal standard in quantitative NMR, GC-MS, and LC-MS assays and for tracer studies requiring deuterium labeling.
  • Deuterium-labeled for tracer and internal standard applications.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Supplied as a solid powder for convenient handling and storage.
  • High chemical purity of 99.7% for reliable analytical performance.
  • Stable when stored under recommended conditions to maintain integrity.