Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isophthalic acid | 121-91-5 | 100.0% | 166.13 | 50 G
SDP

Catalog No. 5000401164
Encompass_Preferred

Isophthalic acid is a biological material or organic compound extensively utilized in life science research.

  • Purity of 99.96%
  • Molecular weight of 166.13
  • Formula C8H6O4
  • CAS number 121-91-5
  • Solid appearance
  • White to off-white color
  • Ships at room temperature
  • Powder storage at -20°C for 3 years, 4°C for 2 years
  • Storage in solvent at -80°C for 6 months, -20°C for 1 month
  • Solubility of 200 mg/mL in DMSO

Catalog No. 50-004-01164 Supplier Medchemexpress LLC Supplier No. HY2132350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isophthalic acid is a biological material or organic compound extensively utilized in life science research.

  • Purity of 99.96%
  • Molecular weight of 166.13
  • Formula C8H6O4
  • CAS number 121-91-5
  • Solid appearance
  • White to off-white color
  • Ships at room temperature
  • Powder storage at -20°C for 3 years, 4°C for 2 years
  • Storage in solvent at -80°C for 6 months, -20°C for 1 month
  • Solubility of 200 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.