Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophthalic acid, 99%

Catalog No. aaa1444530
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444530 250 g
AAA144450B 1000 g
AAA144450I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1444530 Supplier Thermo Scientific Chemicals Supplier No. A1444530
Only null left
Add to Cart

CAS: 121-91-5 | C8H6O4 | 166.132 g/mol

Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.132
MDL Number MFCD00002516
InChI Key QQVIHTHCMHWDBS-UHFFFAOYSA-N
Synonym isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
PubChem CID 8496
ChEBI CHEBI:30802
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 344°C to 348°C (sublimation)
Density 1.54
Quantity 250 g
Beilstein 1909332
Merck Index 14,5197
Solubility Information Insoluble in water.
Formula Weight 166.13
Percent Purity 99%
Chemical Name or Material Isophthalic acid
EINECSNumber 204-506-4
RTECSNumber NT2007000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.