Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isophthaloyl chloride

Catalog No. 503124587 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-4587 100 g
50-312-4588 1 kg
50-312-4562 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-4587 Supplier Sigma Aldrich Supplier No. I19403100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isophthaloyl chloride

Specifications

CAS 99-63-8
Melting Point 43°C to 44°C
Boiling Point 276°C
Molecular Formula C8H4Cl2O2
Linear Formula C6H4-1,3-(COCl)2
Quantity 100 g
MDL Number MFCD00000678
Synonym Isophthaloyl dichloride
Molecular Weight (g/mol) 203.02
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.