Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isophytol | 505-32-8 | MFCD00048380 | 99.7% | 296.53 g/mol | C20H40O | 500 G
SDP

Catalog No. 5000282787
Encompass_Preferred

Isophytol is a branched-chain terpene alcohol used as an organic reagent and intermediate in biochemical and life-science research. It is a colorless to light-yellow liquid commonly used in organic synthesis, assay development, and fragrance/intermediate applications.

  • High purity, 99.7%.
  • Liquid physical form; density ~0.841 g/cm3.
  • Molecular weight 296.53 g/mol.
  • Suitable for organic synthesis and assay development.
  • Available in bulk packaging, including 500 g quantities.

Catalog No. 50-002-82787 Supplier Medchemexpress LLC Supplier No. HYW127745500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isophytol is a branched-chain terpene alcohol used as an organic reagent and intermediate in biochemical and life-science research. It is a colorless to light-yellow liquid commonly used in organic synthesis, assay development, and fragrance/intermediate applications.

  • High purity, 99.7%.
  • Liquid physical form; density ~0.841 g/cm3.
  • Molecular weight 296.53 g/mol.
  • Suitable for organic synthesis and assay development.
  • Available in bulk packaging, including 500 g quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.