Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Isopinocampheylborane TMEDA Complex 98.0+%, TCI America™
SDP

Catalog No. I07965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07965G Supplier TCI America Supplier No. I07965G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 68297-74-5
Molecular Formula C26H50B2N2
Molecular Weight (g/mol) 412.32
MDL Number MFCD00013212
InChI Key YIPGSNPACSKQKJ-AUDPXGPISA-N
Synonym (+)-N,N′C-Bis(monoisopinocampheylborane)-N,N,N′C,N′C-tetramethylethylenediamine
PubChem CID 12482036
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine;[(1R,3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]boron
SMILES [B]C1CC2CC(C1C)C2(C)C.[B]C1CC2CC(C1C)C2(C)C.CN(C)CCN(C)C

Specifications

Melting Point 141°C
Color White
Quantity 5 g
UN Number 1325
Formula Weight 416.35
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Isopinocampheylborane TMEDA Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.