Learn More
MilliporeSigma Isopore Membrane Filter, 100/pk
Supplier:  MilliporeSigma TETP01300
Encompass
8.0 um pore size, hydrophilic polycarbonate membrane, 13 mm diameter. The Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy.
Catalog No. NC0506814