Learn More
Selleck Chemical LLC Isoprenaline (Isoproterenol) HCl S2566-50mg
SDP
Catalog No. 503108864
Encompass_Preferred
Isoprenaline (Isoproterenol) HCl is a non-selective beta-adrenergic receptor agonist used for the treatment of bradycardia and heart block
Catalog No. 50-310-8864 Supplier Selleck Chemical LLC Supplier No. S256650MG
Isoprenaline (Isoproterenol) HCl is a non-selective beta-adrenergic receptor agonist used for the treatment of bradycardia and heart block