Learn More
Medchemexpress LLC Isoprenaline hydrochloride | 51-30-9 | 99.9% | 100 MG
SDP
Catalog No. 5000407805
Encompass_Preferred
Isoprenaline (Isoproterenol) hydrochloride is a non-selective, orally active β-adrenergic receptor agonist. It exhibits potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities. This compound is suitable for research related to bradycardia and bronchial asthma.
  • Non-selective β-adrenergic receptor agonist
  • Orally active
  • Potent peripheral vasodilator
  • Potent bronchodilator
  • Potent cardiac stimulant
  • Used in research for bradycardia and bronchial asthma
Catalog No. 50-004-07805 Supplier Medchemexpress LLC Supplier No. HYB0468100MG
Isoprenaline (Isoproterenol) hydrochloride is a non-selective, orally active β-adrenergic receptor agonist. It exhibits potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities. This compound is suitable for research related to bradycardia and bronchial asthma.
  • Non-selective β-adrenergic receptor agonist
  • Orally active
  • Potent peripheral vasodilator
  • Potent bronchodilator
  • Potent cardiac stimulant
  • Used in research for bradycardia and bronchial asthma