Learn More
Medchemexpress LLC Isoprenaline hydrochloride (Isoproterenol hydrochloride) | 51-30-9 | 99.9% | 1 ML
SDP
Catalog No. 5000408215
Encompass_Preferred
Isoprenaline hydrochloride is a non-selective, orally active β-adrenergic receptor agonist. It possesses potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities. It can be utilized for research into bradycardia and bronchial asthma.
  • Non-selective β-adrenergic receptor agonist
  • Orally active
  • Potent peripheral vasodilator
  • Potent bronchodilator
  • Potent cardiac stimulating activities
  • Research applications: useful for studying conditions such as bradycardia and bronchial asthma
Catalog No. 50-004-08215 Supplier Medchemexpress LLC Supplier No. HYB046810MM/1ML
Isoprenaline hydrochloride is a non-selective, orally active β-adrenergic receptor agonist. It possesses potent peripheral vasodilator, bronchodilator, and cardiac stimulating activities. It can be utilized for research into bradycardia and bronchial asthma.
  • Non-selective β-adrenergic receptor agonist
  • Orally active
  • Potent peripheral vasodilator
  • Potent bronchodilator
  • Potent cardiac stimulating activities
  • Research applications: useful for studying conditions such as bradycardia and bronchial asthma