Learn More
Cayman Chemical Isoprinosine, Purity: ≥98%, CAS: 36703-88-5, 50 mg
SDP
Catalog No. 502972559
Encompass_Preferred
A complex of acetaminobenzoic acid, dimethylaminoisopropanol, and inosine in a 3:3:1 ratio; has a number of immunomodulatory effects, including inducing T-lymphocyte differentiation, augmenting macrophage and NK cell functions, and stimulating IL-2 production; shows antiviral activity
Catalog No. 50-297-2559 Supplier Cayman Chemical Supplier No. 1649250 MG
A complex of acetaminobenzoic acid, dimethylaminoisopropanol, and inosine in a 3:3:1 ratio; has a number of immunomodulatory effects, including inducing T-lymphocyte differentiation, augmenting macrophage and NK cell functions, and stimulating IL-2 production; shows antiviral activity