Learn More
TARGETMOL CHEMICALS INC Isopropamide Iodide 100MG
SDP
Catalog No. 502879777
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Isopropamide iodide is a long-acting anticholinergic drug used for peptic ulcers and other gastrointestinal disorders, and as a muscarinic receptor antagonist for diseases affecting cognition [including Alzheimer's disease]. Purity 99.91%
Catalog No. 50-287-9777 Supplier TARGETMOL CHEMICALS INC Supplier No. T5051100MG
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Isopropamide iodide is a long-acting anticholinergic drug used for peptic ulcers and other gastrointestinal disorders, and as a muscarinic receptor antagonist for diseases affecting cognition [including Alzheimer's disease]. Purity 99.91%