Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropamide (iodide) | 71-81-8 | 98.0% | 1 ML
SDP

Catalog No. 5000413667
Encompass_Preferred

Isopropamide iodide is a long-acting quaternary anticholinergic agent used in peptic ulcer and other gastrointestinal disorders marked by hyperacidity and hypermotility. This product is provided as a 10 mM solution in DMSO, suitable for various laboratory applications.

  • Long-acting anticholinergic agent
  • Supports research in gastrointestinal disorders
  • Helps manage hyperacidity and hypermotility
  • Sealed storage away from moisture
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent

Catalog No. 50-004-13667 Supplier Medchemexpress LLC Supplier No. HYB166710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropamide iodide is a long-acting quaternary anticholinergic agent used in peptic ulcer and other gastrointestinal disorders marked by hyperacidity and hypermotility. This product is provided as a 10 mM solution in DMSO, suitable for various laboratory applications.

  • Long-acting anticholinergic agent
  • Supports research in gastrointestinal disorders
  • Helps manage hyperacidity and hypermotility
  • Sealed storage away from moisture
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.