Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropan(ol-d) 98+ atom% D, Thermo Scientific™

Catalog No. AC174800250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC174800250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC174800250 Supplier Thermo Scientific Chemicals Supplier No. 174800250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3979-51-9
Molecular Formula C3H8O
Molecular Weight (g/mol) 61.102
InChI Key KFZMGEQAYNKOFK-QYKNYGDISA-N
Synonym propan-2-2h ol, 2-propan ol-d, 2-propanol-d, 2-propanol-o-d, 2-propanol-d1, isopropyl alcoh ol-d, o-2h-2-propanol, propan-2-2 h ol, 2-propanol-od, 98 atom % d, 2-propan ol-d 98 atom % d
PubChem CID 520960
IUPAC Name 2-deuteriooxypropane
SMILES CC(C)O

Specifications

Physical Form Liquid
Color Colorless
Quantity 25 g
Density 0.798
Deuterium (D) 98.0% min.
Water 1% max. (Coulometric)
Linear Formula (CH3)2CHOD
Formula Weight 61.1
Chemical Name or Material Isopropan(ol-d), 98+ atom % D
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.