Learn More
Andwin Scientific BDH Isopropyl Alcohol 99%, 67-63-0, MFCD00011674, 1L
SDP
Catalog No. NC1637718
Encompass_Preferred
Formula: (CH₃)₂CHOH, MW: 60.1 g/mol, Boiling Pt: 82 °C (1013 hPa), Melting Pt: –89 °C; BDH Isopropyl Alcohol (Aka 2-Propanol) is suitable for general laboratory use including cytology histology organic synthesis cleaning prep scale synthesis. Packaged In 1-Liter Plastic Bottles
Catalog No. NC1637718 Supplier Andwin Scientific Supplier No. BDH11331LP
Formula: (CH₃)₂CHOH, MW: 60.1 g/mol, Boiling Pt: 82 °C (1013 hPa), Melting Pt: –89 °C; BDH Isopropyl Alcohol (Aka 2-Propanol) is suitable for general laboratory use including cytology histology organic synthesis cleaning prep scale synthesis. Packaged In 1-Liter Plastic Bottles