Learn More
VWR INTERNATIONAL LLC 2-Propanol ≥99% for biotechnology, 1l
Catalog No. NC9704454
Encompass
2-Propanol ≥99% for biotechnology, 67-63-0, MFCD00011674, (CH₃)₂CHOH, 60.1 g/mol, 1l
Catalog No. NC9704454 Supplier VWR INTERNATIONAL LLC Supplier No. 97064700
2-Propanol ≥99% for biotechnology, 67-63-0, MFCD00011674, (CH₃)₂CHOH, 60.1 g/mol, 1l