Learn More
Sigma Aldrich Fine Chemicals Biosciences Isopropanol 70 in H2O1L
Catalog No. 502898254
Encompass
The product is a 70% SOLUTIONon of isopropanol (i-PrOH 2-propanol) in water. i-PrOH is a renewable secondary alcohol that can be utilized as a potential substitute to petroleum-derived gasoline. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range 277-346K. Its photocatalytic degradation has been examined by various methods.
Catalog No. 50-289-8254 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5639351L
The product is a 70% SOLUTIONon of isopropanol (i-PrOH 2-propanol) in water. i-PrOH is a renewable secondary alcohol that can be utilized as a potential substitute to petroleum-derived gasoline. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range 277-346K. Its photocatalytic degradation has been examined by various methods.