Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99.9% (HPLC Grade), Fisher BioReagents™

Catalog No. BP26324 Shop All Fisherbrand Products
Change view
Click to view available options
Packaging:
EcoSafPak™ Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
BP26324 EcoSafPak™ Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP26324

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Submicron Filtered

  • Suitable for precipitating nucleic acids
  • Submicron filtered
  • UV absorbance is measured and reported
  • Meets ACS specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance 0.025 AU max. at 254nm, 0.13 AU max. at 230nm, 0.25 AU max. at 220nm
Color Colorless
Density 0.785g/cm³
Identification Pass Test
Packaging EcoSafPak™ Amber Glass
Physical Form Liquid
Vapor Pressure 43mmHg at 20°C
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa/s at 20°C
Water 0.10% max.
Boiling Point 81°C
Formula Weight 60.1
Percent Purity ≥99.9%
Grade Also meets ACS specifications
Chemical Name or Material Isopropanol, 99.9%
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.