Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol 99, TARR

Catalog No. 502985616
Encompass
Change view
Click to view available options
Quantity:
4 x 1 gal.
350 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5616 4 x 1 gal.
50-298-5649 350 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-298-5616 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T12468
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0

Specifications

Quantity 4 x 1 gal.
Boiling Point 180.1 °F
Molecular Weight (g/mol) 60.1 g/mool
Percent Purity ≥99.5%
Grade Semiconductor
Chemical Name or Material Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.