Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol 99, TARR

Catalog No. 502985649
Encompass
Change view
Click to view available options
Quantity:
4 x 1 gal.
350 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5649 350 lb.
50-298-5616 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-298-5649 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T10688
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0

Specifications

Quantity 350 lb.
Packaging Drum
Molecular Formula C3H8O
Boiling Point 180.1 °F
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.10
Grade Semiconductor
Chemical Name or Material Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.