Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99%, Vion Biosciences
SCI_ED

Catalog No. S02496 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 L
3.8 L
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02496 1 L
S02495 3.8 L
S02494 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S02496 Supplier Aldon Corporation Supplier No. VNIX02351L
Add to Cart
Add to Cart

This 99% isopropanol is an excellent alternative to ethanol as a dehydrant and during paraffinization as an substitute for solvents such as chloroform, xylene, and benzene.

  • This 99% isopropanol is an excellent alternative to ethanol as a dehydrant and during paraffinization as an substitute for solvents such as chloroform, xylene, and benzene.

Chemical Identifiers

Molecular Formula (CH3)2CHOH
Molecular Weight (g/mol) 60.1 g/mol
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym 2-propanol, isopropanol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH Data not available.
Melting Point -90°C
Color Colorless
CAS Max % 100%
Appearance Clear, colorless liquid.
Decomposition Information Data not available.
Density 0.786 to 0.790 (at 20°C)
Quantity 1 L
Explosion Limit 2%
CAS 67-63-0
Flash Point 12°C
Identification Isopropanol
Ignition Point 399°C
Infrared Spectrum Yes
Odor Aromatic odor.
Packaging Natural Plastic Cylinder
Physical Form Liquid
Product Line Vion Histology
Assay Percent Range 100%
Specific Gravity 0.0786 to 0.790 (at 20°C)
Linear Formula (CH3)2CHOH
Vapor Pressure 33 mm (at 20°C)
UN Number UN1219
Water 0%
Concentration 99%
Formulation 95% Isopropanol
Solubility Information Complete in water.
Boiling Point 82°C
Formula Weight 60.1 g/mol
Percent Purity 99%
Grade Histological
Concentration or Composition (by Analyte or Components) 99% Isopropanol
Chemical Name or Material Isopropanol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Causes serious eye irritation.
Hazard Statement May cause drowsiness or dizziness.
ShelfLife 3 Years
DOTInformation UN1219, Isopropanol, 3(II), Limited quantity ≤ 1 L
EINECSNumber 200-661-7
RTECSNumber NT8050000
Recommended Storage Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.