Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOI00204C1 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOI00204C1 4 L Safety-coated Glass Bottle
RSOI00201C 1 L Amber Glass Bottle
RSOI00204C 4 L Amber Glass Bottle
RSOI002020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RSOI00204C1 Supplier Ricca Chemical Company Supplier No. RSOI00204C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For General Use

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
Quantity 4 L
Packaging Safety-coated Glass Bottle
Physical Form Liquid
Residue after Evaporation max. 0.001%
Water max. 0.2%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 82.5°C
Molecular Weight (g/mol) 60.096
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Isopropanol
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.