Learn More
Crescent Chemical Co Inc Isopropanol, 67-63-0, MFCD00011674, 250mL
SDP
Catalog No. 50248388
Encompass_Preferred
Molecular Formula C3H8O, Molecular Weight 60.095, Molecular Biology Grade, Suitable for the precipitation of nucleic acids. When compared to ethanol 50 % less is required for nucleic acid precipitation. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-388 Supplier Crescent Chemical Co Inc Supplier No. SE39559.01
Molecular Formula C3H8O, Molecular Weight 60.095, Molecular Biology Grade, Suitable for the precipitation of nucleic acids. When compared to ethanol 50 % less is required for nucleic acid precipitation. Storage Conditions: +15 °C TO +30 °C