Learn More
Crescent Chemical Co Inc ISOPROPANOL MOL BIO GRADE
SDP
Catalog No. 50248389
Encompass_Preferred
Isopropanol molecular biology grade Suitable for the precipitation of nucleic acids. When compared to ethanol 50 % less is required for nucleic acid precipitation. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 39559.02 CA
Catalog No. 50-248-389 Supplier Crescent Chemical Co Inc Supplier No. SE39559.02
Isopropanol molecular biology grade Suitable for the precipitation of nucleic acids. When compared to ethanol 50 % less is required for nucleic acid precipitation. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 39559.02 CA