Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99+%, specified according to the requirements of Ph.Eur.

Catalog No. AC396980025
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC396980025 2.5 L
AC396982500 250 mL
AC396980010 1 L
AC396980100 10 L
AC396980250 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC396980025 Supplier Thermo Scientific Chemicals Supplier No. 396980025
  • $114.50
    $97.65 / Each of 1

    Save $16.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 67-63-0 | C3H8O | 60.096 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Absorbance 0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm
Color Brown to Purple
Appearance Clear colorless liquid
Density 0.7850g/mL
Quantity 2.5 L
Flash Point 12°C
Identification C: Passes Test (EP)
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Low Melting Solid
Refractive Index 1.3760 to 1.3780
Specific Gravity 0.785
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa.s (20°C)
Water 0.5% max.
Boiling Point 81.0°C to 83.0°C
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Formula Weight 60.1
Percent Purity ≥99%
Chemical Name or Material Isopropanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness
Causes serious eye irritation
Highly flammable liquid and vapor
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray
Wear protective gloves/ protective clothing/face protection
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Keep away from heat/sparks/open flames/hot surfaces. - No smoking
Ground/Bond container and receiving equipment

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-661-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.