Learn More
Sigma Aldrich Fine Chemicals Biosciences Isopropenyl acetate produc25KG
Catalog No. 502907155
Encompass
Isopropenyl acetate is a superior acetylating agent and decomposes into ketene and acetone by heating in presence of acid catalysts. Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols phenols and amines under solvent free conditions.
Catalog No. 50-290-7155 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1091425KG
Isopropenyl acetate is a superior acetylating agent and decomposes into ketene and acetone by heating in presence of acid catalysts. Iodine in isopropenyl acetate is a unique catalyst for the acetylation of a variety of alcohols phenols and amines under solvent free conditions.