Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropenylboronic Acid Pinacol Ester

Catalog No. 6632125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-321-25G 5 g
66-321-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 66-321-25G Supplier Sigma Aldrich Supplier No. 6632125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used forPalladium-catalyzed Suzuki-Miyaura cross-coupling reactions Inverse-electron-demand Diels-Alder reaction Simmons-Smith Cyclopropanation Reaction Polyene cyclization Stereoselective aldol reactions Grubbs cross-metathesis reaction Intramolecular Suzuki-Miyaura reaction Stereoselective cross-metathesis Dipolar cycloadditionIodosulfonylation Asymmetric conjugate addition and intramolecular hydroacylation Reagent used in preparation of various therapeutic kinase and enzymatic inhibitors

Specifications

CAS 126726-62-3
Density 0.894 g/mL at 25°C
Boiling Point 47°C to 49°C9, mbar
Molecular Formula C9H17BO2
Refractive Index n20/D 1.4320
Linear Formula C9H17BO2
MDL Number MFCD08276843
Quantity 5 g
Synonym 2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4,4,5,5-Tetramethyl-2-(isopropenyl)-1,3,2-dioxaborolane; 4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane; 4,4,5,5-tetramethyl-2-(1-methylethenyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol) 168.04 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.