Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl 1,1-dimethyl-1,2,3,6-tetrahydroazepino[4,5-b]indole-5-carboxylate | 942145-77-9 | 98.0% | C18H22N2O2 | 250 MG
SDP

Catalog No. 5000358590
Encompass_Preferred

Isopropyl 1,1-dimethyl-1,2,3,6-tetrahydroazepino[4,5-b]indole-5-carboxylate is a laboratory chemical used for the manufacture of substances. This product is intended for research use only by experienced personnel.

  • Appearance: Light yellow to yellow solid
  • Purity: 98.0%
  • Store in a cool, well-ventilated area, away from direct sunlight and ignition sources
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent storage: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-58590 Supplier Medchemexpress LLC Supplier No. 250MGHY50110
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl 1,1-dimethyl-1,2,3,6-tetrahydroazepino[4,5-b]indole-5-carboxylate is a laboratory chemical used for the manufacture of substances. This product is intended for research use only by experienced personnel.

  • Appearance: Light yellow to yellow solid
  • Purity: 98.0%
  • Store in a cool, well-ventilated area, away from direct sunlight and ignition sources
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent storage: -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.