Learn More
Medchemexpress LLC Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-alpha-D-glucopyranoside | 40592-88-9 | MFCD28053519 | 99.9% | 389.40 g/mol | C17H27NO9 | 1 ML
SDP
Catalog No. 5000240417
Encompass_Preferred
TLR4-IN-C34 is a small-molecule, orally active inhibitor of Toll-like receptor 4 (TLR4) used in preclinical research to modulate innate immune signaling. It has been shown to reduce systemic inflammation in animal models of endotoxemia and necrotizing enterocolitis and is provided in formats suitable for both in vitro and in vivo studies.
  • Orally active TLR4 inhibitor used in preclinical inflammation studies.
  • Demonstrated to reduce systemic inflammation in endotoxemia and necrotizing enterocolitis models.
  • Available as a solid and as a 10 mM solution in DMSO for convenient dosing.
  • High purity (99.9%), suitable for research applications.
  • Molecular formula C17H27NO9 and molecular weight 389.40 g/mol.
  • Store powder at -20°C and store solutions at -80°C to maximize stability.
Catalog No. 50-002-40417 Supplier Medchemexpress LLC Supplier No. HY10757510MM 1ML
TLR4-IN-C34 is a small-molecule, orally active inhibitor of Toll-like receptor 4 (TLR4) used in preclinical research to modulate innate immune signaling. It has been shown to reduce systemic inflammation in animal models of endotoxemia and necrotizing enterocolitis and is provided in formats suitable for both in vitro and in vivo studies.
  • Orally active TLR4 inhibitor used in preclinical inflammation studies.
  • Demonstrated to reduce systemic inflammation in endotoxemia and necrotizing enterocolitis models.
  • Available as a solid and as a 10 mM solution in DMSO for convenient dosing.
  • High purity (99.9%), suitable for research applications.
  • Molecular formula C17H27NO9 and molecular weight 389.40 g/mol.
  • Store powder at -20°C and store solutions at -80°C to maximize stability.