Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl 2-bromo-2-methylpropanoate, 97%, Thermo Scientific™

Catalog No. SB01764EE
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01764EE 50 g
SB01764EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB01764EE Supplier Thermo Scientific Chemicals Supplier No. SB01764EE
Add to Cart

Chemical Identifiers

CAS 51368-55-9
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00055235
InChI Key UNZJYKKJZGIFCG-UHFFFAOYSA-N
Synonym isopropyl 2-bromo-2-methylpropanoate, isopropyl 2-bromo-2-methylpropionate, 2-bromo-2-methylpropionic acid isopropyl ester, isopropyl 2-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, 1-methylethyl ester, isopropyl 2-bromo-2-methyl-propanoate, methylethyl 2-bromo-2-methylpropanoate, 2-bromo-iso-butyric acid isopropyl ester, propan-2-yl 2-bromanyl-2-methyl-propanoate, 2-bromoisobutyric acid isopropyl ester
PubChem CID 2796293
IUPAC Name propan-2-yl 2-bromo-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)Br

Specifications

CAS Min % 97.0
Density 1.2600g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula (CH3)2CBrCO2CH(CH3)2
Quantity 50 g
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material Isopropyl 2-bromo-2-methylpropanoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.