Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl 2-Bromo-2-methylpropionate 97.0+%, TCI America™
SDP

Catalog No. I092025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I092025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I092025G Supplier TCI America Supplier No. I092025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51368-55-9
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00055235
InChI Key UNZJYKKJZGIFCG-UHFFFAOYSA-N
Synonym isopropyl 2-bromo-2-methylpropanoate, isopropyl 2-bromo-2-methylpropionate, 2-bromo-2-methylpropionic acid isopropyl ester, isopropyl 2-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, 1-methylethyl ester, isopropyl 2-bromo-2-methyl-propanoate, methylethyl 2-bromo-2-methylpropanoate, 2-bromo-iso-butyric acid isopropyl ester, propan-2-yl 2-bromanyl-2-methyl-propanoate, 2-bromoisobutyric acid isopropyl ester
PubChem CID 2796293
IUPAC Name propan-2-yl 2-bromo-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)Br

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 3272
Formula Weight 209.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl 2-Bromo-2-methylpropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.