Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl 4-Aminobenzoate 98.0+%, TCI America™
SDP

Catalog No. A027325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A027325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A027325G Supplier TCI America Supplier No. A027325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18144-43-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017108
InChI Key JWCPZKNBPMSYND-UHFFFAOYSA-N
Synonym isopropyl 4-aminobenzoate, 4-amino-benzoic acid isopropyl ester, isopropyl p-aminobenzoate, 4-aminobenzoic acid isopropyl ester, benzoic acid, 4-amino-, 1-methylethyl ester, benzoic acid, p-amino-, isopropyl ester, methylethyl 4-aminobenzoate, isopropyl-4-aminobenzoate, 4-isopropoxycarbonylaniline, acmc-1brt8
PubChem CID 87470
IUPAC Name propan-2-yl 4-aminobenzoate
SMILES CC(C)OC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 25 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropyl 4-Aminobenzoate
EINECSNumber (3)-1443
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.