Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Acetate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. p-7214150 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
3 x 1.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3006 3 x 1.2 mL
11-102-3048 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-3006 Supplier MilliporeSigma Supelco Supplier No. PHR21093X1.2ML
Only null left
Add to Cart
Add to Cart

Pharmaceutical secondary standards for applications in quality control. Provides pharma laboratories and manufacturers with a convenient, cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 108-21-4
Melting Point -73°C (literature)
Density 0.872 g/mL (at 25°C (literature))
Boiling Point 85°C to 91°C (literature)
Flash Point 4°C (Not applicable)
Molecular Formula C5H10O2
Linear Formula CH3COOCH(CH3)2
Refractive Index n20/D 1.377 (literature)
MDL Number MFCD00008877
Quantity 3 x 1.2 mL
Synonym Isopropyl Acetate; Acetic Acid Isopropyl Ester
Vapor Density 3.5 (vs air)
Vapor Pressure 47 mmHg (20°C)
Formula Weight 102.13
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.