Learn More
Sigma Aldrich Fine Chemicals Biosciences Isopropyl acetate puriss.1L
Catalog No. 502899768
Encompass
Isopropyl acetate is an isopropyl ester of acetic acid. It participates in the mesoporous Al-MCM-41 (Si/Al 55 and 104) and Al Zn-MCM-41 (Si/(AlZn)52) molecular sieves catalyzed alkylation of m-cresol. It is widely used for the incorporation of aroma to various cosmetics and food products. Vapor-liquid equilibria for its binary mixture with CO2 at higher pressures has been evaluated.
Catalog No. 50-289-9768 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 459601LF
Isopropyl acetate is an isopropyl ester of acetic acid. It participates in the mesoporous Al-MCM-41 (Si/Al 55 and 104) and Al Zn-MCM-41 (Si/(AlZn)52) molecular sieves catalyzed alkylation of m-cresol. It is widely used for the incorporation of aroma to various cosmetics and food products. Vapor-liquid equilibria for its binary mixture with CO2 at higher pressures has been evaluated.