Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Acetate, Puriss p.a., ≥99.5% (GC), Solstice

Catalog No. 60048472 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-472 250 mL Glass bottle
60-048-471 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-472 Supplier Solstice Supplier No. 45960250ML
Add to Cart
Add to Cart

Puriss. p.a.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Density 0.872g/mL at 25°C
Boiling Point 85°C
Explosion Limit 1.8% (37°F), 8%
Flash Point 4°C (39.2°F)
Ignition Point 894°F
Packaging Glass bottle
Refractive Index 1.377
Linear Formula CH3COOCH(CH3)2
Quantity 250 mL
UN Number UN1220
Beilstein 1740761
Vapor Density 3.5 (vs air)
Vapor Pressure 47mmHg (20°C)
Formula Weight 102.13g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Isopropyl acetate
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation class 3 - PG 2 - Isopropyl acetate
RTECSNumber 203-561-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.