Learn More
Description
- Process histological and cytological specimens with the help of this histological grade of isopropyl alcohol
- Available in a 55 gallon container
- Minimizes over-dehydration in smaller biopsies
- Excellent for lipid extraction, allowing for increased paraffin infiltration
- Excellent dye solvent for special stains
Specifications
Specifications
| Chemical Name or Material | Isopropyl Alcohol, 2-Propanol, IPA, Isopropanol, Propan-2-ol |
| CAS | 67-63-0 |
| Packaging | Bottle |
| Boiling Point | 81°C to 83°C (177.8°F to 181.4°F) at 760 mmHg |
| Percent Purity | 99% |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.